Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heavy Duty AC Powered Xenon Stroboscopes, Nidec Shimpo

Supplier: NIDEC SHIMPO AMERICA
DT-311A-230V DT-311A
The DT-311A stroboscope provides the illusion of “stopped motion” by adjusting the strobe’s flash rate, equipment appear to be standing still or in slow motion.

  • Aluminum construction provides exceptional durability
  • High polish and focused reflector eliminates blind viewing areas
  • External trigger allows unit to be synchronized with equipment
  • High accuracy of 0.01% of FS reading is ideal for QC inspection

The DT-311A measures rotational (RPM) or reciprocating (strokes per minute) speeds with the same precision as an electronic digital tachometer. Ideally suited for printing inspection, motion analysis, preventive maintenance and speed measurement of repetitive motion, DT-311A strobes provide timely data on vibrating parts, leaks, spray patterns and a variety of industrial and scientific operations the DT-311A stroboscope is extremely easy to maneuver and operate via its lightweight and compact design and simplified user interface. Its rugged metal construction provides unequaled reliability in the harshest environments.



 

ORDER

Expand All / Collapse All
Description Range Accuracy Resolution Supplier No. VWR Catalog Number Unit Price Quantity
Heavy Duty, AC Powered Xenon Stroboscope 40-35,000 FPM 0.01% of FS 40-5,000 = 0.1 FPM 5,001-8,000 = 0.2FPM 8,001-10,000 = 0.5FPM 10,001-35,000 = 1FPM DT-311A 75836-584 Each Retrieving
Heavy Duty, AC Powered Xenon Stroboscope, 230V 40-35,000 FPM 0.01% of FS 40-5,000 = 0.1 FPM 5,001-8,000 = 0.2FPM 8,001-10,000 = 0.5FPM 10,001-35,000 = 1FPM DT-311A-230V 75836-586 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty