Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dual 2-in-1 Hand Held Stroboscope and Tachometers, Nidec Shimpo

Supplier: NIDEC SHIMPO AMERICA
ST-5000
The ST-5000 combination stroboscope/tachometer portable velocity analyzing and measuring device is ideal for machinery process inspection.

  • Combination Stroboscope & Tachometer combines two separate products
  • Quick mode selection provides operations between Stroboscopes and Tachometer
  • Low power draw LEDs along with rechargeable battery provides long operation time
  • Process speed detected by tachometer instantly taken to sctroboscope flash rate

The 2 in 1 unit eliminates the need to purchase and carry around two separate test instruments, plus streamlines time to analyze process machinery. The LED stroboscope functionality of the ST-5000 is capable of flashing its light in a synchronous frequency to the operating machinery, creating the illusion of viewing slowed or frozen images. Utilizing this phenomenon of slowing or stopping the motion with the adjusted flash rate of the ST-5000’s LED lights, machine parts and processes may be inspected for defects, aiding in preventative maintenance programs. The added adjustable flash duration feature enables a clearer stopped image and is ideal for monitoring repetitive processes such as on high-speed printing lines.

Ordering information: Includes 4 AA Rechargeable Batteries, Reflective Tape, and Battery Recharger.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Dual 2-in-1 LED Stroboscope & Tachometer ST-5000 75836-580 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Accuracy 0.013% of FS
Resolution 1,000 = 0.1 , 1,000 = 1
Range 15-200,000 FPM

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty