Imaging Multimeter with IGM™, Extech
Supplier: Extech
DM284
75839-486EA 971.22 USD
75839-486
Imaging Multimeter with IGM™, Extech
The FLIR DM284 is an industrial-grade imaging multimeter built for durability to ensure longevity in rugged environments.
  • Speeds up troubleshooting
The thermal imaging function helps pinpoint electrical problems associated with,‎ for example,‎ electrical panels,‎ electrical connectors,‎ and wires,‎ and speeds up troubleshooting by visually guiding the user to the precise location without making contact with the target equipment. Black color.
Certifications: UL,‎ CE,‎ FCC,‎ CB.
Ordering information: Complete wth? TA82 premium test leads,? TA70 alligator clips,? TA60 thermocouple probe with adapter,? TA84 test lead holder storage/tripod accessory,? 4 × AAA Energizer batteries,? English-only quick start guide.