You Searched For: NMR Spectrometers
Nuclear magnetic resonance, or NMR, is a research technique used in spectroscopy that exploits the magnetic properties of atomic nuclei. NMR spectroscopy determines the physical and chemical properties of atoms or the molecules in which they are contained. Spectrometers available from VWR feature high-sensitivity and can be used as stand-alone tools. NMR spectroscopy is used to research organic molecules, although it can be used in a wide range of applications, for any samples containing nuclei with spin.
Nuclear magnetic resonance, or NMR, is a research technique used in spectroscopy that exploits the magnetic properties of atomic nuclei. NMR spectroscopy determines the physical and chemical properties of atoms or the molecules in which they are contained. Spectrometers available from VWR feature high-sensitivity and can be used as stand-alone tools. NMR spectroscopy is used to research organic molecules, although it can be used in a wide range of applications, for any samples containing nuclei with spin.
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: NANALYSIS CORP.
Description: The Nanalysis 60 MHz benchtop NMR Spectrometers are compact 60 MHz, 1.4 T, all-in-one spectrometers that are both easy-to-use and easy-to-site whereve...