Penicillin : Streptomycin : Amphotericin B solution Cell Culture Reagent 100X
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
091674049
IC1674049EA 57.02 USD
IC1674049
Penicillin : Streptomycin : Amphotericin B solution Cell Culture Reagent 100X
Penicillin : Streptomycin : Amphotericin B
Penicllin-Streptomycin-Amphotericin B is used to supplement cell culture media.
  • Penicillin Concentration:10,000 IU/mL
  • Streptomycin Concentration:10 mg/mL
  • Amphotericin Concentration: 25 μg/mL
  • Effective against yeast, fungi, gram-positive and gram-negative bacteria
Penicillin acts by inhibiting bacterial cell-wall synthesis
Recommended working concentration is 10 mg/L
Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome and causes miscoding. Amphotericin B interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out. Antimicrobial spectrum: Gram-negative bacteria, Gram-positive bacteria, fungi and yeasts.