Kovac's indole reagent for microbiology

Supplier: NORTHEAST LAB SERVICES MS
Danger

Synonyms: 4-(Dimethylamino)benzaldehyde solution

X1150
101374-994EA 78.98 USD
101374-994
Kovac's indole reagent for microbiology
Kovac's indole reagent

Kovac's Reagent is ideal for use with Indole tests.


  • Amber Bottle
  • Black Cap
  • Eye Dropper on the Side
  • Fill Volume: 25 mL


Contains 4-(Dimethylamino) benzaldehyde.

Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00003381
CAS Number: 100-10-7
UN: 2920
ADR: 8 (3),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR