Lauryl Sulfate Broth, Tubes, Northeast Lab Services
Supplier: Northeast Lab Services
T1486 T1485
101374-600PK 102.24 USD
101374-600 101374-598
Lauryl Sulfate Broth, Tubes, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Lauryl Sulfate Broth is a prepared medium that includes a handy Durham tube.
  • Durham Tube Included
  • Glass Construction
  • Screw Cap
  • Different Sizes Available
  • Fill Volume: 10 mL
The broth features glass construction and secure screw caps.
It's available in multiple sizes.