Endo Agar, Plates, Northeast Lab Services

Supplier: Northeast Lab Services
P1700
101374-174PK 49.18 USD
101374-174
Endo Agar, Plates, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Endo Agar is contained in a standard petri dish.

  • Fill Volume: 20 mL
  • Prepared Medium
  • Standard Petri Dish
  • Size: 15 mm x 100 mm

The prepared medium has a fill volume of 20 mL.

The petri dish measures 100 mm x 15 mm.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR