Potato Dextrose Agar, Plates, Northeast Lab Services

Supplier: Northeast Lab Services
P1510
101374-076PK 64.25 USD
101374-076
Potato Dextrose Agar, Plates, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Potato Dextrose Agar offers a low pH and deep-fill medium.

  • Fill Volume: 25 mL
  • Deep-Fill
  • Low pH
  • Prepared Medium
  • Size: 15 mm x 100 mm

The prepared medium has a fill volume of 20 mL.

It is contained in a standard petri dish measuring 15 mm x 100 mm.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR