CIN (Yersinia) Agar, Plates, Northeast Lab Services

Supplier: Northeast Lab Services
P1160
101373-950PK 86.52 USD
101373-950
CIN (Yersinia) Agar, Plates, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
CIN Agar is a plated, prepared medium.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Size: 15 mm x 100 mm
  • Prepared Medium

It has a fill volume of 20 mL.

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR