Tryptic Soy Broth, Tubes, Northeast Lab Services
Supplier: Northeast Lab Services
T2013 T2020
101374-910PK 31.33 USD
101374-910 101374-898
Tryptic Soy Broth, Tubes, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Available in glass and autoclavable plastic, Tryptic Soy Broth is a flexible, useful culture in any lab.
  • Variety of Sizes
  • Paragon
  • Lecithin and Polysorbate Available
  • Autoclavable Plastic and Glass
Tryptic Soy Broth is available with a number of media formulations.
NEL produces liquid and agar media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.