innuPREP DYEpure Kit, Analytik Jena
Supplier: Analytik Jena
innuPREP
845-KS-5020050 845-KS-5020050 845-KS-5020250 845-KS-5020250
10755-618EA 588.61 USD
CA10755-616 10755-618 CA10755-618 10755-616
innuPREP DYEpure Kit, Analytik Jena
Nucleic Acid Reagents Sanger Sequencing Reagents
The innuPREP DYEpure Kit is an especially effective, fast tool for removing fluorescence-labeled dideoxynucleoside triphosphates (dye terminators) from sequencing reactions.
  • Effective removal of dye terminators from sequencing reactions
  • Novel two-step technology based on spin filter columns results in a very simple procedure
  • Extremely fast – full purification in just 5 minutes
  • Elution volume reduced down to 10µl
The innuPREP DYEpure Kit is an especially effective, fast tool for removing fluorescence-labeled dideoxynucleoside triphosphates (dye terminators) from sequencing reactions. Dye terminators are typically used for sequencing by chain termination and can interfere with the sequence during read-out. Patented Dual Chemistry (DC) technology allows researchers to omit unnecessary wash steps and time-consuming precipitation with ethanol. This reduces the process to just binding and elution, allowing the entire purification process to be completed in no more than 5 minutes. Thanks to the high quality of the eluate and its exceptional purity, the innuPREP DYEpure Kit provides ideal preparation for subsequent sequencing runs.