Would you like to log in again?
innuPREP DNA Mini Kit, Analytik Jena
Supplier: Analytik Jena US
845-KS-1040250 845-KS-1040050
The universal innuPREP DNA Mini Kit has been specially designed for fast, efficient purification of genomic DNA from a variety of starting materials.
  • Flexible and universally applicable for a broad range of starting materials
  • Patented DC technology for faster lysis and highly efficient gDNA binding
  • Spin filter columns used for isolation
  • CE-IVD certified
The universal innuPREP DNA Mini Kit has been specially designed for fast, efficient purification of genomic DNA from a variety of starting materials. The kit utilizes patented Dual Chemistry (DC) technology, which combines a stringent lysis buffer with a novel binding buffer to help minimize the time required to purify DNA. The result is an extraction process that takes no more than 8 minutes (not including lysis). Spin filter columns are at the heart of the isolation process, which is very easy to perform and offers a binding capacity of up to 100 µg of gDNA. The innuPREP DNA Mini Kit is also certified for in-vitro-diagnostics use (CE-IVD).
ORDER
Expand All / Collapse All
Description No. of Reactions Supplier No. VWR Catalog Number Unit Price Quantity
innuPREP DNA Mini Kit 250 Reaction 250 845-KS-1040250 10755-586 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
innuPREP DNA Mini Kit 50 Reaction 50 845-KS-1040050 10755-584 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Contents Tissue samples of up to 50 mg Rodent tail specimens 0.5 – 1 cm in length Paraffin samples (tissue) Eukaryotic cells (max. 5 × 10^6) Buccal swabs
Time Approx. 8 minutes after lysis
Purity 1.7 – 2.0
Binding Capacity Column binding capacity: 100 µg gDNA
Yield Depending on the type and quantity of the starting material, up to 65 µg
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty