innuPREP Blood DNA Mini Kit, Analytik Jena
Supplier: Analytik Jena
innuPREP
845-KS-1020250 845-KS-1020050 845-KS-1020250 845-KS-1020050
10755-576EA 181.87 USD
10755-576 10755-578 CA10755-576 CA10755-578
innuPREP Blood DNA Mini Kit, Analytik Jena
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The innuPREP Blood DNA Mini Kit is a highly efficient tool for directly isolating DNA from whole blood samples of up to 400μl.
  • Fast, direct isolation of genomic DNA from whole blood samples of up to 400μl
  • High yields of up to 30μg and extremely high-quality gDNA, depending on the sample and the amount used
  • CE-IVD certified
  • 2 protocols: < 200μl and up to 400μl blood samples
  • Tested for EDTA and citrate stabilized and for fresh or frozen blood samples (including long-term storage)
  • Based on patented low-salt DC chemistry
The innuPREP Blood DNA Mini Kit is a highly efficient tool for directly isolating DNA from whole blood samples of up to 400μl
The innuPREP Blood DNA Mini Kit is extremely fast and easy to use. Isolation of genomic DNA, free of inhibitors or impurities, can be completed in just 24 minutes. The kit is also CE-IVD certified and has already undergone successful testing: genomic DNA has been isolated from whole blood samples and then used in subsequent diagnostic applications.
This kit guarantees extremely high-quality DNA that can be used immediately for photometric determinations, PCR, or other downstream applications.
Specifications
Starting material: Whole blood samples (up to 400 µl) - Fresh or frozen blood; Stabilizers: EDTA or citrate
Column binding capacity: > 50 µg gDNA
Average purity: 1.7 – 2.0
Extraction time: Approx. 24 minutes, including lysis
Average yield: Depends on sample and the used volume; up to 30 µg gDNA