innuCONVERT Bisulfite Basic Kit, Analytik Jena
Supplier: Analytik Jena
innuCONVERT
845-IC-1000080 845-IC-1000080 845-IC-1000040
10755-554EA 401.83 USD
10755-554 CA10755-554 CA10755-744
innuCONVERT Bisulfite Basic Kit, Analytik Jena
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The innuCONVERT Bisulfite Basic Kit allows users to completely convert non-methylated cytosine to uracil in just a few hours
  • Complete conversion of non-methylated cytosine to uracil in just a few hours
  • Flexible for use in all commonly used thermocyclers or heating blocks
  • Denaturation and conversion combined in a single work sequence
The innuCONVERT Bisulfite Basic Kit allows users to completely convert non-methylated cytosine to uracil in just a few hours. DNA sample denaturation and bisulfite treatment are combined in the same reaction vessel. After a total reaction time of approx. 3 hours, the converted DNA is isolated and desulfonated in a column. The eluted DNA is then ready to use in downstream applications (such as PCR, sequencing, etc).
DNA sample denaturation and bisulfite treatment are combined in the same reaction vessel.