PhiKan083, BioVision
Supplier: BioVision
9440-5 9440-25
75838-240EA 729.68 USD
75838-240 75838-238
PhiKan083, BioVision
Protein Expression Protein Expression Reagents
PhiKan083 is a p53 stabilizing agent.
  • Preferentially binds mutated (Y220C) p53 over wild-type p53 at a site distinct from functional DNA/protein interaction regions
  • Stabilizing agent
  • Slows rate of thermal denaturation of p53-Y220C
Preferentially binds mutated (Y220C) p53 over wild-type p53 at a site distinct from functional DNA/protein interaction regions. PhiKan083 slows rate of thermal denaturation of p53-Y220C.