GC Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M434-500G M434-2.5KG
89225-518EA 583.98 USD
CA95021-824 89225-518
GC Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and cultivation of gonococci.
  • Microbiological culture media
  • Supplied in poly bottles
Requires added blood or hemoglobin and other supplements.