Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Sharps Container Systems, Bracket Compatible, Multiple Colors

Supplier: VWR International
VWR®
8708V 8708TV 8708CV 8708TYV
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps.

  • Stackable 5.4 Quart Containers
  • Containers Feature Biohazard Labeling, Biohazard Symbol, Locking Lid, and Counterbalanced Door
  • Available in an Assortment of Colors
  • Autoclavable

The stackable containers measure 5²⁄₅ qt.

Available in an assortment of colors, the autoclavable containers feature Biohazard labeling, a Biohazard symbol, and a counterbalanced door.



 

ORDER

Expand All / Collapse All
Capacity Dimensions Color VWR Catalog Number Unit Price Quantity
5.4 qt. (5.1 L) 26.7 x 8.9 x 26.7 cm (10.5 x 3.5 x 10.5") Opaque Red 75800-274 Case of 12 Retrieving
5.4 qt. (5.1 L) 26.7 x 8.9 x 26.7 cm (10.5 x 3.5 x 10.5") Translucent Red 75800-272 Case of 12 Retrieving
5.4 qt. (5.1 L) 26.7 x 8.9 x 26.7 cm (10.5 x 3.5 x 10.5") Translucent Yellow 75800-278 Case of 12 Retrieving
5.4 qt. (5.1 L) 26.7 x 8.9 x 26.7 cm (10.5 x 3.5 x 10.5") Clear 75800-276 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty