EZBlock™ Protease Inhibitor Cocktail XIII, EDTA & Animal-Free, BioVision
Supplier: BioVision
AB287911-5VIAL
75837-934EA 583.92 USD
75837-934
EZBlock™ Protease Inhibitor Cocktail XIII, EDTA & Animal-Free, BioVision
Cell Culture Supplements and Additives
A cocktail of six broad specificity protease inhibitors to be used for mammalian cell/tissue extracts as well as plant and bacterial cell extracts.
This cocktail is EDTA-free and Animal-free.