TAS-301

Synonyms: 3-(Bis(4-methoxyphenyl)methylene)indolin-2-one

9644-10 9644-50
75837-776EA 145.93 USD
75837-776 75837-778
TAS-301
TAS-301

TAS-301 is an inhibitor of smooth muscle cell migration and proliferation.


Blocks voltage-independent calcium influx and downstream PKC signaling. Inhibits neointimal thickening after balloon catheter injury to the rat common carotid artery. Inhibits calcium-dependent signal transduction and cytoskeletal reorganization.

Formula: C₂₃H₁₉NO₃
MW: 357.4 g/mol
MDL Number: MFCD05865245
CAS Number: 193620-69-8
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR