L-778123 hydrochloride

9638-10 9638-50
75837-900EA 181.76 USD
75837-900 75837-902
L-778123 hydrochloride
L-778123 hydrochloride

L-778123 hydrochloride is a potent inhibitor of FPTase and GGPTase-I with an IC₅₀ of 2 nM and 98 nM, respectively, in enzyme inhibition determination.


 

Formula: C₂₂H₂₀ClN₅O
MW: 405.88 g/mol
CAS Number: 253863-00-2
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR