JH-II-127

Synonyms: (4-((5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl)amino)-3-methoxyphenyl)(morpholino)methanone

9632-5 9632-25
75837-698EA 145.93 USD
75837-698 75837-700
JH-II-127
JH-II-127

JH-II-127 is a highly potent, selective, and cell-permeable LRRK2 inhibitor, with IC₅₀ of 6.6 nM, 2.2 nM ,47.7 nM for LRRK2-wild-type, LRRK2-G2019S, LRRK2-A2016T.


JH-II-127 substantially inhibits Ser910 and Ser935 phosphorylation of both wild-type LRRK2 and G2019S mutant at a concentration of 0.1-03 μM in a variety of cell types and is capable of inhibiting Ser935 phosphorylation in mouse brain following oral delivery of doses as low as 30 mg/kg.

Formula: C₁₉H₂₁ClN₆O₃
MW: 416.87 g/mol
MDL Number: MFCD28975160
CAS Number: 1700693-08-8
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR