GDC-0941 Dimethanesulfonate

9618-10 9618-50
75837-530EA 157.38 USD
75837-530 75837-532
GDC-0941 Dimethanesulfonate
GDC-0941 Dimethanesulfonate

GDC-0941 is a potent, selective, inhibitor of pan-class I PI3 kinase (PI3K), with IC₅₀ values of 3, 33, 3, 75, 1230 and 580 nM for p110 α, β, δ and γ isoforms, DNA-PK and mTOR respectively.


Displays antineoplastic activity.

Formula: C₂₅H₃₅N₇O₉S₄
MW: 705.85 g/mol
MDL Number: MFCD21608527
CAS Number: 957054-33-0
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR