Radotinib

Synonyms: 4-Methyl-N-(3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl)-3-((4-(pyrazin-2-yl)pyrimidin-2-yl)amino)benzamide

9613-50 9613-10
75837-842EA 171.34 USD
75837-842 75837-844
Radotinib
Radotinib

Radotinib is a BCR-ABL1-specific second-generation tyrosine kinase inhibitor (TKI).


Also inhibits PDGFR thereby blocking PDGFR-mediated signal transduction pathways. Displays antineoplastic activities.

Formula: C₂₇H₂₁F₃N₈O
MW: 530.5 g/mol
MDL Number: MFCD27956910
CAS Number: 926037-48-1
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR