AZD-3759

Synonyms: (R)-4-((3-Chloro-2-fluorophenyl)amino)-7-methoxyquinazolin-6-yl 2,4-dimethylpiperazine-1-carboxylate

9609-5 9609-25
75837-508EA 624.61 USD
75837-508 75837-506
AZD-3759
AZD-3759

AZD-3759 is a potent epidermal growth factor receptor (EGFR) inhibitor, with potential antineoplastic activity.


AZD-3759 binds to and inhibits the activity of EGFR as well as certain mutant forms of EGFR. This prevents EGFR-mediated signaling, and may lead to both induction of cell death and inhibition of tumor growth in EGFR-overexpressing cells. 

Formula: C₂₂H₂₃ClFN₅O₃
MW: 459.9 g/mol
MDL Number: MFCD29058564
CAS Number: 1626387-80-1
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR