Bedaquiline

Supplier: BioVision

AB285381-5MG AB285381-25MG
75837-232EA 1253.61 USD
75837-232 75837-230
Bedaquiline
Bedaquiline

Bedaquiline is an anti-tuberculosis drug.


Bedaquiline's unique and specific anti-mycobacterial activity derives from inhibition of the proton pump of mycobacterial ATP synthase, which is a critical enzyme in the ATP synthesis of M. tuberculosis. Binding of bedaquiline to the oligomeric and proteolipic subunit-c of mycobacterial ATP synthase leads to inhibition of ATP synthesis, which subsequently results in bacterial death.

Formula: C₃₂H₃₁BrN₂O₂
MW: 555.5 g/mol
MDL Number: MFCD22628872
CAS Number: 843663-66-1
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR