Ellipticine

Synonyms: 5,11-Dimethyl-6H-pyrido[4,3-b]carbazole

9579-25 9579-5
75837-438EA 577.91 USD
75837-438 75837-436
Ellipticine
Ellipticine

Ellipticine is a naturally occurring plant alkaloid that acts as a DNA intercalating agent and DNA topoisomerase II inhibitor.


Ellipticine is also shown to block p53 protein phosphorylation.

Formula: C₁₇H₁₄N₂
MW: 246.3 g/mol
MDL Number: MFCD00010524
CAS Number: 519-23-3
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR