IRAK-1/4 Inhibitor

Synonyms: N-(1-(2-Morpholinoethyl)-1H-benzo[d]imidazol-2-yl)-3-nitrobenzamide

9578-25 9578-5
75837-434EA 577.91 USD
75837-434 75837-432
IRAK-1/4 Inhibitor
IRAK-1/4 Inhibitor

IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor associated kinase (IRAK) with IC₅₀ values of 0.3 μM and 0.2 μM for IRAK-1 and IRAK-4, respectively.


It displays IC₅₀ values >10 µM in a panel of 27 other kinases tested.

Formula: C₂₀H₂₁N₅O
MW: 395.41 g/mol
MDL Number: MFCD09752602
CAS Number: 509093-47-4
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR