FIIN-2

9577-5 9577-25
75837-430EA 624.61 USD
75837-430 75837-428
FIIN-2
FIIN-2

FIIN-2 is a potent, selective, irreversible, next-generation covalent FGFR inhibitor.


FIIN-2 is the first inhibitor that can potently inhibit the proliferation of cells dependent upon the gatekeeper mutants of FGFR1 or FGFR2, which confer resistance to first-generation clinical FGFR inhibitors such as NVP-BGJ398 and AZD4547.

Formula: C₃₅H₃₈N₈O₄
MW: 634.73 g/mol
MDL Number: MFCD28386553
CAS Number: 1633044-56-0
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR