Droxinostat

9570-5 9570-25
75837-396EA 531.21 USD
75837-396 75837-394
Droxinostat
Droxinostat

Droxinostat is a selective inhibitor of HDAC3, HDAC6, and HDAC8 with IC₅₀ values of 16.9, 2.47 and 1.46 μM, respectively.


Droxinostat sensitizes cells to apoptosis by decreasing c-FLIPL and c-FLIPS expression, reducing cell survival, and inducing apoptosis.

Formula: C₁₁H₁₄ClNO₃
MW: 243.69 g/mol
MDL Number: MFCD01326592
CAS Number: 99873-43-5
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR