Avibactam Sodium

Supplier: BioVision

AB285380-25 AB285380-5
75837-390EA 403.42 USD
75837-390 75837-392
Avibactam Sodium
Avibactam Sodium

Avibactam (NXL104) is a potent inhibitor of class A and C serine β-lactamases, including KPC carbapenemases.


NXL104 quantitatively inhibits BlaC, the only chromosomally encoded β-lactamase from Mycobacterium tuberculosis, by forming a carbamyl adduct with the enzyme.

Formula: C₇H₁₀N₃NaO₆S
MW: 287.222 g/mol
MDL Number: MFCD28900719
CAS Number: 1192491-61-4
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR