SB-742457

Synonyms: 3-(Phenylsulphonyl)-8-(piperazin-1-yl)quinoline3-(Phenylsulfonyl)-8-(piperazin-1-yl)quinoline

9568-5 9568-25
75837-386EA 134.25 USD
75837-386 75837-388
SB-742457
SB-742457

SB-742457 is a highly selective 5-HT6 receptor antagonist with potential cognition, memory and learning-enhancing effects; displays >100-fold selectivity over other receptors.


In anxiolytic and antidepressant-like activity. SB-742457 shows increasing pro-cognitive effects in patients with Alzheimer’s Disease (AD).  .

Formula: C₁₉H₁₉N₃O₂S
MW: 353.44 g/mol
MDL Number: MFCD12828872
CAS Number: 607742-69-8
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR