H-7 dihydrochloride

9543-25 9543-5
75837-268EA 134.25 USD
75837-268 75837-270
H-7 dihydrochloride
H-7 dihydrochloride

H-7 is a Protein kinase inhibitor that inhibits the activity of PKA, PKC, PKG and MLCK with IC₅₀ values of 3.0, 6.0, 5.8 and 97.0 µM, respectively.

Formula: C₁₄H₁₇N₃O₂S·2HCl
MW: 364.29 g/mol
MDL Number: MFCD00036961
CAS Number: 108930-17-2
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR