17-AAG, EZSolution™

Synonyms: 17-(Allylamino)-17-demethoxygeldanamycin(4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

9495-1
75836-988EA 163.46 USD
75836-988
17-AAG, EZSolution™
17-AAG

A 10 mM (1 mg in 171 µl) of Hsp90 inhibitor 17-AAG (Cat


No.1774) in anhydrous DMSO.

Formula: C₃₁H₄₃N₃O₈
MW: 585.7 g/mol
MDL Number: MFCD04973892
CAS Number: 75747-14-7
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR