BMS-387032 hydrochloride

Supplier: BioVision

Synonyms: SNS-032

AB219467-25MG AB219467-5MG
75836-974EA 388.53 USD
75836-974 75836-976
BMS-387032 hydrochloride
BMS-387032 hydrochloride

SNS-032 is a potent CDK inhibitor.


Inhibits cyclin-dependent kinases (cdks) 9, 2 and 7 with IC₅₀ values of 4, 38 and 62 nM respectively. Displays no activity against 190 additional kinases (IC₅₀ >1000 nM). Prevents tumor cell-induced VEGF secretion and in vitro angiogenesis.

Formula: C₁₇H₂₄N₄O₂S₂·HCl
MW: 417 g/mol
CAS Number: 345627-90-9
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR