Palbociclib isethionate

Synonyms: 6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 2-hydroxyethanesulfonate6-Acetyl-8-cyclopentyl-5-methyl-2-((5-(piperazin-1-yl)pyridin-2-yl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 2-hydroxyethanesulphonate

9480-25 9480-5
75836-940EA 531.21 USD
75836-940 75836-938
Palbociclib isethionate
Palbociclib isethionate

PD0332991 is a highly specific inhibitor of Cdk4 (IC₅₀ = 11 nM) and Cdk6 (IC₅₀ = 16 nM).


Displays selectivity for Cdk4/6 over a panel of 34 additional protein kinases (IC₅₀ values are > 10 μM)...

Formula: C₂₄H₂₉N₇O₂·C₂H₆O₄S
MW: 573.66 g/mol
MDL Number: MFCD22666589
CAS Number: 827022-33-3
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR