Sodium orthovanadate, activated

Supplier: BioVision

AB301822-5
75837-660EA 197.92 USD
75837-660
Sodium orthovanadate, activated
Sodium orthovanadate

Activated Sodium orthovanadate is a depolymerized vanadate prepared from Sodium orthovanadate (Cat No. 9470) that displays enhanced ability to inhibit endogenous phosphatases in a competitive manner.


  • Preserves the phosphorylation state in cells and cell lysates.

Formula: Na₃O₄V
MW: 183.91 g/mol
MDL Number: MFCD00003511
CAS Number: 13721-39-6
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR