Bromocriptine mesylate

2916-50 2916-250
75836-822EA 88.63 USD
75836-822 75836-824
Bromocriptine mesylate
Bromocriptine mesylate

Bromocriptine is a potent Cytochrome P450 3A inhibitor (~770 nM, see cat#K702).


It also acts as a dopamine receptor agonist via the D2 receptor (Ki = 2.5 nM). As a result, it is used in combination therapy for Parkinsons disease.

Formula: C₃₂H₄₀BrN₅O₅ · CH₃SO₃H
MDL Number: MFCD00069218
CAS Number: 22260-51-1
UN: 3316

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR