Adhesive Vinyl GHS OSHA Labels 'Blank GHS Label AMAT Format', HCL Labels
Supplier: HCL Labels
GHS-1
103050-834EA 3.31 USD
103050-834
Adhesive Vinyl GHS OSHA Labels 'Blank GHS Label AMAT Format', HCL Labels
Labels
This write-in adhesive vinyl label complies with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Blank writing surface
  • Personalize labels easily
This label is compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.