Adhesive Vinyl GHS OSHA Labels 'Sodium Phosphate', HCL Labels
Supplier: HCL Labels
GH11203VN0047 GH21203VN00235 GH41203VN00235 GH21203VN00475 GH31203VN00235 GH11203VN00475 GH41203VN0023 GH11203VN0023 GH31203VN0035 GH21203VN0047 GH31203VN00475 GH41203VN00475 GH21203VN0023 GH11203VN00235 GH21203VN0035 GH41203VN00355 GH11203VN0035 GH41203VN0035 GH31203VN0023 GH11203VN00355 GH41203VN0047 GH31203VN0047 GH21203VN00355 GH31203VN00355
103040-236PK 59.31 USD
103040-236 103040-238 103032-248 103047-620 103032-246 103032-244 103024-228 103024-236 103047-624 103024-226 103024-234 103047-622 103024-232 103047-618 103047-626 103024-230 103047-616 103032-254 103040-240 103032-252 103040-242 103040-232 103032-250 103040-234
Adhesive Vinyl GHS OSHA Labels 'Sodium Phosphate', HCL Labels
Labels
These ready-made laminated adhesive vinyl labels comply with the updated OSHA HazCom secondary container labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS). All sizes are protected with a chemical resistant lamination and a shelf/outdoor life of two years.
Certifications: RoHS compliant.