Adhesive Vinyl GHS OSHA Labels 'Silicon Dioxide', HCL Labels
Supplier: HCL Labels
GH41104VN00235 GH11104VN00475 GH11104VN00235 GH21104VN0035 GH21104VN0047 GH31104VN00475 GH41104VN0035 GH21104VN00475 GH41104VN00475 GH11104VN0047 GH31104VN0023 GH31104VN00235 GH41104VN0023 GH41104VN0047 GH31104VN0035 GH11104VN00355 GH11104VN0035 GH21104VN00235 GH31104VN00355 GH31104VN0047 GH21104VN0023 GH41104VN00355 GH11104VN0023 GH21104VN00355
103032-128PK 74.8 USD
103032-128 103047-510 103032-126 103047-500 103032-124 103047-504 103047-502 103024-188 103047-508 103024-186 103047-506 103024-184 103024-182 103024-192 103040-050 103024-190 103032-122 103040-052 103040-054 103032-120 103032-130 103040-044 103040-046 103040-048
Adhesive Vinyl GHS OSHA Labels 'Silicon Dioxide', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Comply with labeling requirements
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN globally harmonized system for the classification and labeling of chemicals (GHS).
Certifications: RoHS compliant.