Adhesive Vinyl GHS OSHA Labels 'Boric Acid', HCL Labels
Supplier: HCL Labels
GH20434VN0023 GH20434VN00475 GH10434VN0035 GH40434VN00475 GH40434VN00235 GH20434VN00355 GH40434VN00355 GH40434VN0035 GH10434VN0047 GH30434VN0023 GH40434VN0023 GH30434VN0035 GH40434VN0047 GH20434VN00235 GH20434VN0047 GH10434VN00235 GH10434VN00355 GH30434VN00355 GH30434VN00235 GH10434VN0023 GH20434VN0035 GH30434VN00475 GH10434VN00475 GH30434VN0047
103021-138PK 59.31 USD
103021-138 103028-992 103028-990 103021-134 103044-890 103021-136 103028-998 103028-996 103028-994 103036-848 103029-000 103036-846 103036-844 103036-842 103044-892 103021-142 103036-840 103044-894 103036-850 103044-884 103021-144 103044-886 103044-888 103021-140
Adhesive Vinyl GHS OSHA Labels 'Boric Acid', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.