Adhesive Vinyl GHS OSHA Labels 'Antimony', HCL Labels
Supplier: HCL Labels
GH10424VN00235 GH40424VN00475 GH20424VN00355 GH40424VN00355 GH20424VN0023 GH30424VN00355 GH20424VN0047 GH20424VN00235 GH10424VN0023 GH30424VN00475 GH10424VN0035 GH30424VN0035 GH40424VN0023 GH30424VN0047 GH40424VN0047 GH10424VN0047 GH10424VN00475 GH40424VN0035 GH20424VN00475 GH30424VN0023 GH30424VN00235 GH10424VN00355 GH20424VN0035 GH40424VN00235
103028-860PK 39.58 USD
103028-860 103021-018 103044-260 103021-024 103044-262 103021-014 103044-252 103021-016 103028-856 103028-854 103028-864 103028-862 103036-708 103036-706 103036-704 103036-714 103036-712 103036-710 103028-858 103021-020 103044-254 103021-022 103044-256 103044-258
Adhesive Vinyl GHS OSHA Labels 'Antimony', HCL Labels
Labels
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
  • Chemical resistant lamination
  • Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.