Petroleum Sorbent Spill Kit, 3M Company
Supplier: 3M
7000051990
67101-226CS 164.03 USD
67101-226
Petroleum Sorbent Spill Kit, 3M Company
Spill Control Materials
Petroleum folded sorbent (which can be used as booms, pillows or rolls) and high capacity pads.
  • For use in oil-only Sorbent applications.
  • Spill kits combine all the features of 3M Sorbents.
  • Kit is packaged in a 20 gallon drum
This petroleum sorbent spill kit combines the best of both worlds - petroleum folded sorbent (which can be used as booms, pillows or rolls) and high capacity pads. This unique combination allows you to easily select the product to fit the response.