    Sort by:
  • Search Within Results
Optovin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Optovin is a reversible photoactive TRPA1 activator. It specifically activates TRPA1 (EC₅₀ = 2 μM) but not TRPV1 or TRPM8. Optovin activates human TRP...