Dimethyldichlorosilane (DMDCS) Deactivating Agent, Restek
Supplier: Restek
31861
75835-818EA 73.66 USD
75835-818
Dimethyldichlorosilane (DMDCS) Deactivating Agent, Restek
Standards Organic Standards
  • Easy deactivation of liners and other glass surfaces
  • Convenient—25 mL bottle deactivates 60 inlet liners
  • Tested to ensure consistent quality and effectiveness.
Restek offers dimethyldichlorosilane (DMDCS) for deactivating liners and other glassware. DMDCS reacts with active hydroxyl groups on the glass surface to produce a deactivated surface. A simple, step-by-step procedure is included with the product.
Ordering information: Ground transportation shipments only.
Quantity discounts not available.