BAC Resolution Control Standard n-P (6 Components), Restek
Supplier: Restek
36010
75835-432EA 35.89 USD
75835-432
BAC Resolution Control Standard n-P (6 Components), Restek
Standards Organic Standards
The mix is use to verify the retention time for each compound normally included in a blood alcohol test, and to verify that the compounds are resolved from and do not interfere with one another.
  • Includes 1-propanol internal standard
  • Intended for qualitative use only
It consists of acetaldehyde, acetone, ethanol (BAC), methanol, 1-propanol (n-propanol) and 2-propanol (isopropanol).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.