UCMR3 Method 524.3 Standard, Restek
Supplier: Restek
30642
75834-536EA 92.15 USD
75834-536
UCMR3 Method 524.3 Standard, Restek
Standards Organic Standards
This UCMR3 method 524.3 standard contains 9 components: Bromochloromethane 600 μg/ml, Bromomethane (methyl bromide) 2000 μg/ml, 1,3-Butadiene 1000 μg/ml, sec-Butylbenzene 400 μg/ml, Chlorodifluoromethane (CFC-22) 800 μg/ml, Chloromethane (methyl chloride) 2000 μg/ml, 1,1-Dichloroethane 300 μg/ml, n-Propylbenzene 300 μg/ml, 1,2,3-Trichloropropane 300 μg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.