Organonitrogen Pesticide Mix #1 (Rev), Method 525.2 (37 components), Restek
Supplier: Restek
33012
75835-084EA 303.51 USD
75835-084
Organonitrogen Pesticide Mix #1 (Rev), Method 525.2 (37 components), Restek
Standards Organic Standards
Contains: Alachlor, ametryn, atraton, atrazine, bromacil, butachlor, butylate, chlorpropham, cyanazine (bladex), cycloate, diphenamid, EPTC, etridiazole, fenarimol, fluridone (sonar), hexazinone (velpar), metolachlor, metribuzin, MGK-264, molinate, napropamide (devrinol), norflurazon, pebulate, prometon, prometryne, propachlor, propazine, propyzamide, simazine, simetryn, tebuthiuron, terbacil, terbutryn, triadimefon, tricyclazole (beam), trifluralin, vernolate.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.