SOM01.1 VOA Ketone Deuterated Monitoring Compounds, Restek
Supplier: Restek
30625
75834-522EA 289.45 USD
75834-522
SOM01.1 VOA Ketone Deuterated Monitoring Compounds, Restek
Standards Organic Standards
This ketone deuterated monitoring compound contains two components: 2-butanone-d5 and 2-hexanone-d5.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.